Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG1 Rabbit anti-Mouse, Clone: RM106 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB14935
Description
Rabbit monoclonal antibody raised against native mouse IgG1.
- High specificity
- High reactivity
Specifications
| Mouse IgG1 | |
| Monoclonal | |
| Biotin | |
| ELISA (0.005-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| P01868 | |
| Ighg1 | |
| Native purified mouse IgG. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| RM106 | |
| Rabbit monoclonal antibody raised against native mouse IgG1. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| IgG1/Igh-4/VH7183 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 16017 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction