Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2a Kappa Isotype Control (M2AK), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB001288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001288 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001288 Supplier Novus Biologicals Supplier No. NBP196981G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG2a Kappa Monoclonal specifically detects IgG2a Kappa in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG2a Kappa
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone M2AK
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-linked immunosorbent assay
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Purification Method Protein A purified
Quantity 0.5 mL
Primary or Secondary Isotype Control (Primary)
Test Specificity Isotyping assay resulted non-reactive with antisera to mouse IgG1, IgG2b, IgG3, IgA, IgM. Light chain composition has been confirmed by SDS-PAGE.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.