Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2b Isotype Control (MPC-11), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP227231G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227231G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227231G Supplier Novus Biologicals Supplier No. NBP227231G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IgG2b
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MPC-11
Conjugate DyLight 488
Dilution Flow Cytometry 1 ul/1 million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation 50 mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.