Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2b Kappa Light Chain Isotype Control (MG2b), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB001222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001222 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001222 Supplier Novus Biologicals Supplier No. NBP143317APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Control.

Specifications

Antigen IgG2b Kappa Light Chain
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MG2b
Conjugate APC
Dilution Flow Cytometry : 1:10 - 1:1000, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:10-1:500, Control
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mL
Primary or Secondary Isotype Control (Primary)
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.