Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG2b Rabbit anti-Mouse, Clone: RM108 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB14938
Description
Rabbit monoclonal antibody raised against native mouse IgG2b.
- High specificity
- High reactivity
Specifications
Mouse IgG2b | |
Monoclonal | |
Biotin | |
ELISA (0.005-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
P01867 | |
Igh-3 | |
Native purified mouse IgG2b. | |
50 μg | |
Primary | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RM108 | |
Rabbit monoclonal antibody raised against native mouse IgG2b. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
IGHG2B/IgG2b/gamma2b | |
Rabbit | |
Protein A purification | |
RUO | |
16016 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction