Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2c Rabbit anti-Mouse, Clone: RM223, Abnova™

Catalog No. 89035318 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-318 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-318 Supplier Abnova Corporation Supplier No. MAB15110
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against native mouse IgG2c.

  • High specificity
  • High reactivity

Specifications

Antigen Mouse IgG2c
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RM223
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against native mouse IgG2c.
Dilution ELISA (0.01-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-2 ug/mL) Immunoprecipitation Western Blot (nonreduced only) (0.5-2 ug/mL) The optimal worki
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias IGHG2C
Gene Symbols Igh-1b
Host Species Rabbit
Immunogen Native purified IgG2c from mouse hybridoma.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404711
Test Specificity This antibody reacts to the Fc region of mouse IgG2c. No cross reactivity with mouse IgG1, IgG2a, IgG2b, IgG3, IgM, IgA, IgE, human IgG, or rat IgG.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.