Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgG3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB9675100
This item is not returnable.
View return policy
Description
IgG3 Monoclonal specifically detects IgG3 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
IgG3 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects mouse IgG 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with mouse IgG 1 mouse IgG 2A and mouse IgG 2B was observed. | |
Mouse | |
Supernatant |
Western Blot, Flow Cytometry | |
2216B | |
Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
Mouse IgG3 | |
100 μg | |
3502 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction