Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG3 Isotype Control, Alexa Fluor™ 546, Novus Biologicals™
SDP

Catalog No. DDXCM03A546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDXCM03A546 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDXCM03A546 Supplier Novus Biologicals Supplier No. DDXCM03A546
Add to Cart
Add to Cart

Mouse Polyclonal Antibody

IgG3 Polyclonal specifically detects IgG3 in Mouse samples. It is validated for Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence.

Specifications

Antigen IgG3
Applications Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunofluorescence
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Alexa Fluor 546
Dilution Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500
Formulation PBS, 50% glycerol. with No Preservative
Gene Alias DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Gene Symbols IGHG3
Host Species Mouse
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3502
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.