Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgM Antibody Pair, Novus Biologicals™
SDP

Catalog No. NB777439 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB777439 15 Plates
NB777363 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB777439 Supplier R&D Systems Supplier No. NBP34842715PLATES
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody Pair

Mouse IgM Polyclonal antibody specifically detects Mouse IgM in Mouse samples. It is validated for ELISA

Specifications

Antigen Mouse IgM
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Gene Alias Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Quantity 15 Plates
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.