Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgM Kappa Light Chain Isotype Control (MMK), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB001250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001250 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001250 Supplier Novus Biologicals Supplier No. NBP196975AF532

Mouse Monoclonal Antibody

IgM Kappa Light Chain Monoclonal specifically detects IgM Kappa Light Chain in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgM Kappa Light Chain
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone MMK
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page, Fluorophore-linked immunosorbent assay
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Purification Method Multi-step
Quantity 0.5 mL
Primary or Secondary Isotype Control (Primary)
Test Specificity Isotyping assay resulted non-reactive with antisera to mouse IgG1, IgG2a, IgG2b, Ig3, IgA. Light chain composition has been confirmed by SDS-PAGE.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.