Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IGSF2/CD101 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3368A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3368A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3368A Supplier R&D Systems Supplier No. FAB3368A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IGSF2/CD101 Monoclonal specifically detects IGSF2/CD101 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IGSF2/CD101
Applications Flow Cytometry
Classification Monoclonal
Clone 307707
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. A8E0Y8
Gene Alias CD101, CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, Leukocyte Surface Protein, member 2, V7
Gene Symbols CD101
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse IGSF2/CD101 Gln21-Tyr1033 Accession # A8E0Y8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9398
Test Specificity Detects mouse IGSF2/CD101 in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.