Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGSF9 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9140R100UG
This item is not returnable.
View return policy
Description
IGSF9 Monoclonal specifically detects IGSF9 in Mouse samples. It is validated for Flow Cytometry.Specifications
IGSF9 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Dasm1, FP18798, IGSF9A, immunoglobulin superfamily member 9A, immunoglobulin superfamily, member 9, KIAA1355, Nrt1, Protein Turtle Homolog A, TUTLA | |
Mouse myeloma cell line NS0-derived recombinant mouse IGSF9B, Arg21-Leu730, Accession No. Q05BQ1 | |
100 μg | |
57549.0 | |
Mouse | |
Supernatant |
Flow Cytometry | |
993107 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q05BQ1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse IGSF9B in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction