Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IGSF9 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9140S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9140S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9140S100 Supplier R&D Systems Supplier No. FAB9140S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGSF9 Monoclonal specifically detects IGSF9 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IGSF9
Applications Flow Cytometry
Classification Monoclonal
Clone 993107
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q05BQ1
Gene Alias Dasm1, FP18798, IGSF9A, immunoglobulin superfamily member 9A, immunoglobulin superfamily, member 9, KIAA1355, Nrt1, Protein Turtle Homolog A, TUTLA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IGSF9B, Arg21-Leu730, Accession No. Q05BQ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 57549.0
Test Specificity Detects mouse IGSF9B in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.