Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGSF9 Antibody, R&D Systems™

Rat Monoclonal Antibody
$456.00
Specifications
| Antigen | IGSF9 |
|---|---|
| Clone | 993107 |
| Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9140100
![]() |
R&D Systems
MAB9140100 |
100 μg |
Each for $456.00
|
|
|||||
|
MAB9140SP
|
R&D Systems
MAB9140SP |
25 μg | N/A | N/A | N/A | ||||
Description
IGSF9 Monoclonal specifically detects IGSF9 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| IGSF9 | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| Q05BQ1 | |
| 57549 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 993107 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Dasm1, FP18798, IGSF9A, immunoglobulin superfamily member 9A, immunoglobulin superfamily, member 9, KIAA1355, Nrt1, Protein Turtle Homolog A, TUTLA | |
| Mouse myeloma cell line NS0-derived recombinant mouse IGSF9, Arg21-Leu730, Accession # Q05BQ1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IGSF9 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title