Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IKK beta Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB7155
This item is not returnable.
View return policy
Description
IKK beta Monoclonal specifically detects IKK beta in Mouse samples. It is validated for Western Blot.Specifications
| IKK beta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O88351 | |
| IKBKB | |
| E. coli-derived recombinant mouse IKK beta Val530-Asp757 Accession # O88351 | |
| 100 μg | |
| Primary | |
| Detects mouse IKK beta in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 725818 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKB, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3551 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction