Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-1 beta/IL-1F2 Antibody, R&D Systems™
GSA_VAGREENER_CHOICESDP

Catalog No. AF401NA Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF401NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF401NA Supplier R&D Systems Supplier No. AF401NA
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Human∣Mouse∣Pig∣Pseudomonas aeruginosa∣Rabbit∣Rat∣Transgenic Mouse samples. It is validated for Bead-based Immunoassay∣Bioassay∣Cell Culture∣ChIP Assay∣ELISA∣ELISA Capture∣ELISA Standard∣Flow Cytometry∣Immunocytochemistry∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣In vitro Assay∣In vivo Assay∣MUSTag PCR Assay∣Neutralization∣Proximity Ligation Assay∣Simple Western∣Westen Blot∣Western Blot∣Western Blots.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen IL-1 beta/IL-1F2
Applications Bead-based Immunoassay, Bioassay, Cell Culture, ChIP Assay, ELISA, ELISA Capture (Matched Antibody Pair), Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, In vivo Assay, PCR, Neutralization, Proximity Ligation Assay, Simple Western, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization ≤0.25 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. NP_032387
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-1 beta/IL-1F2 Val118-Ser269 Accession # NP_032387
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects mouse IL-1 beta/IL-1F2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Pig, Pseudomonas aeruginosa, Rabbit, Rat, Transgenic Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.