Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-1 beta/IL-1F2 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 4 publications

$549.50 - $2020.00

Specifications

Antigen IL-1 beta/IL-1F2
Clone 166926
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB4011100
GREENER_CHOICE
View Documents
R&D Systems
MAB4011100
100 μg
Each for $549.50
Only null left
Add to Cart
 
MAB4011500
GREENER_CHOICE
View Documents
R&D Systems
MAB4011500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

IL-1 beta/IL-1F2
Western Blot 1 ug/mL
Monoclonal
Supernatant
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
IL1B
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
166926
Western Blot
Unconjugated
Rat
Mouse
P10749
3553
E. coli-derived recombinant mouse IL-1 beta Val118-Ser269 Accession # P10749
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse IL-1 beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, 25-100% cross-reactivity with human, rat, and cotton rat IL-1 beta is obseved and no cross-reactivity with recombinant porcine IL-1 beta , recombinant mouse (rm) IL-1 alpha , or rmIL-18 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.