Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-1 beta/IL-1F2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB401R100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB401R100 100 μg
MAB401RSP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB401R100 Supplier R&D Systems Supplier No. MAB401R100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Mouse samples. It is validated for Neutralization, Immunocytochemistry.

Specifications

Antigen IL-1 beta/IL-1F2
Applications Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 30311R
Conjugate Unconjugated
Dilution Neutralization 1-5 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10749
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols Il1b
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-1 beta/IL-1F2 Val118-Ser269 Accession # P10749
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects mouse IL-1 beta/IL-1F2 in direct ELISAs.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.