Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-1 beta/IL-1F2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB401NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB401NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB401NA Supplier R&D Systems Supplier No. AB401NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 9 publications

IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Mouse samples. It is validated for Western Blot, Simple Western, Neutralization.

Specifications

Antigen IL-1 beta/IL-1F2
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL, Neutralization 2-12 ug/mL
Gene Accession No. P10749
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-1 beta/IL-1F2 Val118-Ser269 Accession # P10749
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects mouse IL-1 beta/IL-1F2 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant cotton rat IL-1 beta and recombinant rhesus monkey IL-1 beta is observed, approximately 15% cross-reactivity with recombinant rat IL-1 beta and recombinant rabbit IL-1 beta is observed, and less than 10% cross-reactivity with recombinant canine IL-1 beta , recombinant equine IL-1 beta , recombinant guinea pig IL-1 beta , recombinant porcine IL-1 beta , recombinant feline IL-1 beta , and recombinant human IL-1 beta is observed.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.