Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-1 beta/IL-1F2 Quantikine HS ELISA Kit

Catalog No. mhslb00
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

From the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

  • A 4.0 hour solid phase ELISA designed to measure mouse IL-1 beta levels in serum and plasma.
  • It contains E. coli-expressed recombinant mouse IL-1 beta and antibodies raised against the recombinant protein.
  • Results obtained using natural IL-1 beta showed linear curves that were parallel to the standard curves obtained using the Quantikine HS kit standards.
  • These results indicate that the Quantikine HS Immunoassay kit can be used to determine relative mass values for natural mouse IL-1 beta.
  • Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested twenty times on one plate to assess intra-assay precision.
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in twenty separate assays to assess inter-assay precision. Assays were performed by at least three technicians.

Specifications

Assay Sensitivity 0.312 pg/mL
Cross Reactivity < 0.5% cross-reactivity observed with available related molecules.< 50% cross-species reactivity observed with species tested.
Product Type ELISA
Specificity Natural and recombinant mouse IL-1 beta
Gene ID (Entrez) 3553
Host Species Mouse
Quantity 1 Kit
Sample Volume Serum (50 μL), EDTA Plasma (50 μL), Heparin Plasma(50 μL)
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Target IL-1 beta/IL-1F2
Total Assay Time 4 hours
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.