Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-1 RI Antibody, R&D Systems™

Hamster Monoclonal Antibody
Supplier: R&D Systems MAB7711SP
This item is not returnable.
View return policy
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Mouse samples. It is validated for Neutralization.Specifications
IL-1 RI | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL1R1 | |
Human cervical epithelial carcinoma cell line HeLa-derived recombinant mouse IL-1 RI | |
25 μg | |
Primary | |
Detects mouse IL-1 RI in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 R3, recombinant mouse (rm) IL-18 R, rmIL-1 RII, recombinant rat IL-1 R6, or rhIL-1 R8 is observed. | |
Mouse | |
Purified |
Neutralization | |
JAMA147 | |
Neutralization 0.05-0.15 ug/mL | |
CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3554 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction