Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-1 RI Fluorescein-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
$174.60 - $339.00
Specifications
Antigen | IL-1 RI |
---|---|
Clone | 129304 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB7712F025
![]() |
R&D Systems
FAB7712F025 |
25 Tests |
Each for $174.60
|
|
|||||
FAB7712F100
![]() |
R&D Systems
FAB7712F100 |
100 Tests |
Each for $339.00
|
|
|||||
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-1 RI | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
IL1R1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse IL-1 RI in direct ELISAs and Western blots. |
129304 | |
Flow Cytometry | |
Fluorescein | |
Rat | |
Mouse | |
P13504 | |
3554 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI Leu20-Lys338 Accession # P13504 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title