Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB417500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB417500 500 μg
MAB417SP 25 μg
MAB417100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB417500 Supplier R&D Systems Supplier No. MAB417500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 36 publications

IL-10 Monoclonal specifically detects IL-10 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-10
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone JES052A5
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.005-0.015 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Accession No. P18893
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Gene Symbols IL10
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-10 Accession # P18893
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3586
Test Specificity Detects mouse IL-10 in sandwich ELISAs and Western blots. In sandwich ELISAs, less than 8% cross-reactivity with recombinant rat IL-10 and 1% cross-reactivity with recombinant cotton rat IL-10 is observed. In sandwich ELISAs, no cross-reactivity with recombinant human (rh) IL-10, recombinant mouse IL-10 R, rhIL-10 R, recombinant canine IL-10, recombinant feline IL-10, or recombinant porcine IL-10 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.