Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-10 R alpha Antibody, R&D Systems™

Hamster Monoclonal Antibody
$208.00 - $679.50
Specifications
Antigen | IL-10R alpha |
---|---|
Clone | 105653 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Mouse samples. It is validated for Western Blot.Specifications
IL-10R alpha | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL10R alpha, IL-10R subunit 1, IL-10R1, IL10RA, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
IL10RA | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
105653 | |
Western Blot | |
Unconjugated | |
Hamster | |
Mouse | |
Q61727 | |
3587 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-10 R alpha Thr24-Asn238 ( Lys188Arg and Glu222Gly) Accession # Q61727 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IL-10 R alpha in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) IL-10 R alpha or rhIL-10 R beta . |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title