Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-11 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF418NA
This item is not returnable.
View return policy
Description
IL-11 Polyclonal specifically detects IL-11 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
IL-11 | |
Polyclonal | |
Unconjugated | |
P47873 | |
IL11 | |
E. coli-derived recombinant mouse IL-11, Gly23-Leu199, Accession # P47873 | |
100 μg | |
Primary | |
Detects mouse IL-11 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.006-0.012 ug/mL | |
Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin | |
Goat | |
Affinity Purified | |
RUO | |
3589 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction