Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-11 R alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF490
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF490 Supplier R&D Systems Supplier No. AF490
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IL-11R alpha Polyclonal specifically detects IL-11R alpha in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-11R alpha
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 2-6 ug/mL
Gene Accession No. Q64385
Gene Alias AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1
Gene Symbols IL11RA
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-11 R alpha Ser22-Gln367 Accession # Q64385
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3590
Test Specificity Detects mouse IL-11 R alpha in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.