Learn More
Description
Specifications
Specifications
| Assay Range | 15.6 - 1,000pg/mL |
| Assay Sensitivity | 10.0pg/mL |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3592 |
| Gene Symbol | IL12A |
| Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-12 p70, Biotin-Conjugate anti-mouse IL-12 p70 monoclonal antibody, Streptavidin-HRP, Mouse IL-12 p70 Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
| Quantity | 96 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
