Learn More
Description
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 1 Kit |
| Species | Mouse |
| Conjugate | Biotin |
| Assay Range | 31.20 to 2000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 16160 |
| Product Line | R&D Systems |
| Packaging Quantity | 5 Plates |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
