Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-12/IL-23 p40 Non Allele-specific DuoSet ELISA

Catalog No. DY2398
Change view
Click to view available options
Packaging Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DY2398 15 Plates
DY239805 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DY2398 Supplier R&D Systems™ Supplier No. DY2398
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse IL-12/IL-23 p40 Non Allele-specific DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-12/IL-23 p40 Non Allele-specific DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-12/IL-23 p40.

Specifications

Product Type ELISA Development Kit
Quantity 1 Kit
Species Mouse
Conjugate Biotin
Assay Range 15.60 to 1000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 16160
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.