Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-12/IL-35 p35 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6688
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6688 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6688 Supplier R&D Systems Supplier No. MAB6688
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen IL-12/IL-35 p35
Applications Immunohistochemistry
Classification Monoclonal
Clone 45806
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_032377
Gene Symbols Il12a
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived mouse IL-12/IL-35 p35 Arg23-Ala215 Accession # NP_032377
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects mouse IL-12/IL-35 p35 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) IL-12 p35+p40 heterodimer or recombinant rat IL-12 p35 is observed. No cross-reactivity with recombinant human IL-12 p35, rmIL-12 p40, rmIL-23 p19+p40 heterodimer, recombinant canine IL-12 p35+p40 heterodimer, or recombinant feline IL-12 p35+p40 heterodimer is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.