Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-12 p70 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB419100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB419100 100 μg
MAB419SP 25 μg
MAB419500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB419100 Supplier R&D Systems Supplier No. MAB419100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 14 publications

IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-12 p70/IL-12A
Applications Western Blot, ELISA
Classification Monoclonal
Clone 48110
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CLMF p35, CLMF1, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL12, IL12 p70, IL-12 subunit p35, IL-12, subunit p35, IL12A, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Gene Symbols IL12A
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-12
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3592
Test Specificity Detects mouse IL-12 in ELISAs and Western blots. Detects an epitope in the p35 subunit. In sandwich ELISAs, does not cross-react with recombinant mouse IL-12 p40.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.