Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Mouse IL-12 p70 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-12 p70/IL-12A.
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 1 Kit |
| Species | Mouse |
| Conjugate | Biotin |
| Assay Range | 39.10 to 2500 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 16159 |
| Packaging Quantity | 5 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
