Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-12 R beta 1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1998
This item is not returnable.
View return policy
Description
IL-12 R beta 1 Polyclonal specifically detects IL-12 R beta 1 in Mouse samples. It is validated for Western Blot.Specifications
IL-12 R beta 1 | |
Polyclonal | |
Unconjugated | |
Q60837 | |
IL12RB1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-12 R beta 1 Val32-Glu561 Accession # Q60837 | |
100 μg | |
Primary | |
Detects mouse IL-12 R beta 1 in direct ELISAs and Western blots. In these formats, approximately 20% cross-reactivity with recombinant human IL-12 R beta 1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454 | |
Goat | |
Affinity Purified | |
RUO | |
3594 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction