Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-12 R beta 2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $526.00
Specifications
Antigen | IL-12 R beta 2 |
---|---|
Clone | 918102 |
Dilution | Blockade of Receptor-ligand Interaction 30-180 ng/mL |
Applications | Blockade of Receptor-ligand Interaction |
Classification | Monoclonal |
Description
IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
IL-12 R beta 2 | |
Blockade of Receptor-ligand Interaction 30-180 ng/mL | |
Monoclonal | |
Rat | |
Mouse | |
P97378 | |
3595 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-12 R beta 2 Met1-Asn637 Accession # P97378 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IL-12 R beta 2 in direct ELISAs. |
918102 | |
Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
Il12rb2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title