Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-13 Antibody, R&D Systems™

Rat Monoclonal Antibody
$227.50 - $526.00
Specifications
Antigen | IL-13 |
---|---|
Clone | 38213R |
Dilution | Western Blot 1μg/mL, ELISA Capture (Matched Antibody Pair) 2-8μg/mL |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB413RSP
![]() |
R&D Systems
MAB413RSP |
25 μg |
Each for $227.50
|
|
|||||
MAB413R100
![]() |
R&D Systems
MAB413R100 |
100 μg |
Each for $526.00
|
|
|||||
Description
IL-13 Monoclonal specifically detects IL-13 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IL-13 | |
Western Blot 1μg/mL, ELISA Capture (Matched Antibody Pair) 2-8μg/mL | |
Monoclonal | |
Supernatant | |
Mouse | |
P20109 | |
3596 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
38213R | |
ELISA, Western Blot | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
E. coli-derived recombinant mouse IL-13 Accession # P20109 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects mouse IL-13 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title