Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB413NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB413NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB413NA Supplier R&D Systems Supplier No. AB413NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-13 Polyclonal specifically detects IL-13 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-13
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 15-30 ug/mL
Gene Accession No. P20109
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Gene Symbols IL13
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-13 Ser26-Phe131 Accession # P20109
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3596
Test Specificity Detects mouse IL-13 in direct ELISAs and Western blots. In direct ELISAs and Westerns, less than 1% cross-reactivity with recombinant human IL-13 is observed. Neutralizes the biological activity of recombinant mouse IL-13, but will not neutralize the biological activity of recombinant human IL-13.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.