Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-13 Recombinant Protein

Catalog No. PIRP87881
Encompass
Click to view available options
Quantity:
100 μg
25 μg
500 μg

E. coli Recombinant Protein

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20109
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation 10 mM sodium phosphate with 150 mM NaCl and no preservative; pH 7.2
Gene ID (Entrez) 16163
Molecular Weight (g/mol) 11.7 kD
Name Mouse IL-13 (aa 26-131)
Quantity 25 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration 0.01 ng/ μg cytokine
Gene Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; Interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Gene Symbol Il13
Specific Reactivity Mouse
Conjugate Unconjugated
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.