Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-15R alpha Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB5511T100UG
This item is not returnable.
View return policy
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-15R alpha | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q60819 | |
IL15RA | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-15 R alpha Gly33-Lys205 Accession # Q60819 | |
RUO | |
3601 | |
Mouse | |
IgG2b |
Flow Cytometry | |
888220 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
Rat | |
100 μg | |
Primary | |
Detects mouse IL-15 R alpha in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction