Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-15R alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $655.00
Specifications
Antigen | IL-15R alpha |
---|---|
Clone | 146018 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Mouse samples. It is validated for Western Blot.Specifications
IL-15R alpha | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
IL15RA | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
146018 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q60819 | |
3601 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-15 R alpha Gly33-Lys205 Accession # Q60819 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IL-15 R alpha in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) IL-2 R alpha , recombinant mouse (rm) IL-2 R beta , rmIL-2 R gamma , or rhIL-15 R alpha . |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title