Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17/IL-17A Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 85 publications
$227.50 - $2070.00
Specifications
Antigen | IL-17/IL-17A |
---|---|
Clone | 50104 |
Dilution | Western Blot 1 ug/mL, Neutralization 0.05-0.15 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
IL-17/IL-17A | |
Western Blot 1 ug/mL, Neutralization 0.05-0.15 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
Q62386 | |
3605 | |
E. coli-derived recombinant mouse IL-17 Thr22-Ala158 Accession # Q62386 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IL-17 in direct ELISAs and Western blots. In direct ELISAs, 100% reactivity with recombinant rat IL-17A and approximately 40% reactivity with recombinant mouse (rm) IL-17A/IL-17F heterodimer is observed. No cross-reactivity with recombinant human IL-17, recombinant canine IL-17, rmIL-17B, rmIL-17C, rmIL-17D, rmIL-17E, or rmIL-17F is observed. |
50104 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
IL17A | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title