Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17B Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1709
This item is not returnable.
View return policy
Description
IL-17B Polyclonal specifically detects IL-17B in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
IL-17B | |
Polyclonal | |
Unconjugated | |
Q9QXT6 | |
IL17B | |
E. coli-derived recombinant mouse IL-17B His21-Phe180 Accession # Q9QXT6 | |
100 μg | |
Primary | |
Detects mouse IL-17B in ELISAs and Western blots. In sandwich immunoassays, approximately 40% cross-reactivity with recombinant human IL-17B is observed and less than 0.3% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17C, rmIL-17D, rmIL-17E, and rmIL-17F is observed. | |
Mouse | |
IgG |
Western Blot, Blockade of Receptor-ligand Interaction, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
Goat | |
Affinity Purified | |
RUO | |
27190 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction