Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17C Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC23061R100UG
This item is not returnable.
View return policy
Description
IL-17C Monoclonal specifically detects IL-17C in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-17C | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_665833 | |
IL17C | |
E. coli-derived recombinant mouse IL-17C His15-Gln194 Accession # NP_665833 | |
RUO | |
27189 | |
Mouse | |
IgG2a |
Flow Cytometry | |
311522 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CX2, Cytokine CX2, IL17C, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
Rat | |
100 μg | |
Primary | |
Detects mouse IL-17C in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction