Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17C Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2306
This item is not returnable.
View return policy
Description
IL-17C Polyclonal specifically detects IL-17C in Mouse samples. It is validated for Western Blot.Specifications
IL-17C | |
Polyclonal | |
Unconjugated | |
NP_665833 | |
IL17C | |
E. coli-derived recombinant mouse IL-17C His15-Gln194 Accession # NP_665833 | |
100 μg | |
Primary | |
Detects mouse IL-17C in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human IL-17C is observed, and less than 5% cross-reactivity with recombinant mouse (rm) IL-17D and rmIL-17F is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CX2, Cytokine CX2, IL17C, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
Goat | |
Affinity Purified | |
RUO | |
27189 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction