Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17C Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB23061
This item is not returnable.
View return policy
Description
IL-17C Monoclonal specifically detects IL-17C in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
IL-17C | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_665833 | |
IL17C | |
E. coli-derived recombinant mouse IL-17C His15-Gln194 Accession # NP_665833 | |
100 μg | |
Primary | |
Detects mouse IL-17C in direct ELISAs. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
311522 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CX2, Cytokine CX2, IL17C, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27189 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction