Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17D Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC2274R100UG
This item is not returnable.
View return policy
Description
IL-17D Monoclonal specifically detects IL-17D in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-17D | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_665836 | |
| IL17D | |
| E. coli-derived recombinant mouse IL-17D Ala25-Arg205 Accession # NP_665836 | |
| RUO | |
| 53342 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 312724 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FLJ30846, IL17D, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-17D in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-17D, recombinant mouse (rm) IL-17, rmIL-17B, rmIL-17C, rmIL-17E, or rmIL-17F is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction