Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17D Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | IL-17D |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Description
IL-17D Polyclonal specifically detects IL-17D in Mouse samples. It is validated for Western Blot, ELISA.Specifications
IL-17D | |
Western Blot 0.1 ug/mL, ELISA | |
Polyclonal | |
Goat | |
Mouse | |
FLJ30846, IL17D, IL-17Dinterleukin 27, IL-22, IL-27interleukin-17D, IL27interleukin-27, interleukin 17D, Interleukin-27 | |
IL17D | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Q8K4C4 | |
53342 | |
E. coli-derived recombinant mouse IL-17D Ala25-Arg205 Accession # Q8K4C4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IL-17D in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human (rh) IL-17D is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17B, rmIL-17C, rmIL-17E, and rmIL-17F is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title