Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17E/IL-25 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1399
This item is not returnable.
View return policy
Description
IL-17E/IL-25 Polyclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot.Specifications
IL-17E/IL-25 | |
Polyclonal | |
Unconjugated | |
NP_542767 | |
IL25 | |
E. coli-derived recombinant mouse IL-17E/IL-25 Val17-Ala169 Accession # NP_542767 | |
100 μg | |
Primary | |
Detects mouse IL-17E/IL-25 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) IL-17E and recombinant rat IL-17E is observed, and less than 1% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17B, rhIL-17C, rhIL-17D, and rhIL-17F is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
Goat | |
Affinity Purified | |
RUO | |
64806 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction