Learn More
Mouse IL-17E/IL-25 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 2 publications
Supplier:  R&D Systems AF1399SP
 View more versions of this product
Catalog No. AF1399SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
IL-17E/IL-25 Polyclonal specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
IL-17E/IL-25
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Goat
Affinity Purified
RUO
64806
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL
NP_542767
IL25
E. coli-derived recombinant mouse IL-17E/IL-25 Val17-Ala169 Accession # NP_542767
25 μg
Primary
Detects mouse IL-17E/IL-25 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) IL-17E and recombinant rat IL-17E is observed, and less than 1% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17B, rhIL-17C, rhIL-17D, and rhIL-17F is observed.
Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only