Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-17F Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC20571G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC20571G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC20571G025 Supplier R&D Systems Supplier No. IC20571G025
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-17F Monoclonal specifically detects IL-17F in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-17F
Applications Flow Cytometry
Classification Monoclonal
Clone 1058A
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q7TNI7
Gene Alias Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F
Gene Symbols Il17f
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse IL-17F Ala21-Val153 Accession # Q7TNI7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112744
Test Specificity Detects mouse IL-17F in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.