Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17F Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2057
This item is not returnable.
View return policy
Description
IL-17F Polyclonal specifically detects IL-17F in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-17F | |
Polyclonal | |
Biotin | |
NP_665855 | |
IL17F | |
E. coli-derived recombinant mouse IL-17F (R&D Systems, Catalog # 2057-IL) Arg29-Ala153 Accession # NP_665855 | |
50 μg | |
Primary | |
Detects mouse IL-17F in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human IL-17F, recombinant mouse (rm) IL-17, rmIL-17B, rmIL-17C, rmIL-17D, and rmIL-17E is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F | |
Goat | |
Affinity Purified | |
RUO | |
112744 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction